Learn More
Polyethylene glycol dimethacrylate, M.W. 8,000, Thermo Scientific Chemicals
533.00 SEK - 5935.00 SEK
Chemical Identifiers
CAS | 25852-47-5 |
---|---|
MDL Number | MFCD00081877 |
Synonym | ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15467355
|
Thermo Scientific Alfa Aesar
046985.MD |
250 mg | N/A | ||||||
15442817
|
Thermo Scientific Alfa Aesar
046985.03 |
1 g |
1515.00 SEK
1g |
In Stock
Log in to see stock. |
|||||
15452817
|
Thermo Scientific Alfa Aesar
046985.06 |
5 g |
5935.00 SEK
5g |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
Description
Polyethylene glycol dimethacrylate is used as adhesives, coatings, sealants, photo resists, solder masks and photopolymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25852-47-5 | |
MFCD00081877 | |
ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate |
Specifications
White | |
Granular Solid | |
MFCD00081877 | |
Soluble in water. | |
Polyethylene glycol dimethacrylate, M.W. 8,000 |
25852-47-5 | |
CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3 | |
ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate | |
Light and moisture sensitive |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.