Learn More
BRAND™ Polypropylene 120° Y-shape Tubing Connector
Three arms at equal angles of 120°
Brand: BRAND™ 153020
Additional Details : Weight : 2.44000kg
Description
Specifications
Polypropylene | |
20 | |
Gray |
65 mm | |
120° connectors |