Learn More
DWK Life Sciences Kimble Chase™ Polypropylene Closure with Ultrasonically Welded PTFE Liner
For use with screw-thread culture and centrifuge tubes
Brand: DWK Life Sciences 73808-13415
5510.00 SEK valid until 2024-10-31
Use promo code "21661" to get your promotional price.
This item is not returnable.
View return policy
Description
- Provides superior closure system capable of maintaining its liner through multiple autoclavable operations
- PTFE-coated surface of exposed liner is chemical-resistant and compatible with sensitive, corrosive and volatile solvents
Specifications
13-415 | |
Screw Cap | |
Polypropylene | |
EPA Water Analysis Vials, Screw Thread Culture and Centrifuge Tubes |
PTFE, Rubber | |
Autoclavable | |
For repeated autoclaving, to provide superior closure system |