Learn More
Technical Service Consultants™ Polypropylene Swab
Brand:  Technical Service Consultants™ TS/19-M
Product Code. 11330945
This item is not returnable. View return policy
Description
Description
  • Extensive range of swabs available with a choice of swab shafts including white birchwood, polypropylene (flexible) and polystyrene (breakable)
  • Tips are formed from pure cotton, alginate and cotton
  • Gamma irradiated to ensure a SAL of at least 10-6
Specifications
Specifications
White birchwood, Polypropylene (flexible) and Polystyrene (breakable)
Cotton
500Pack
Swab
Sterile
Viscose/Dacron
500Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Technical Service Consultants™ Polypropylene Swab > Tip Type: Viscose/Dacron