Learn More
Technical Service Consultants™ Polystyrene L-shaped Spreader
Use Technical Service Consultants™ Polystyrene L-shaped Spreader for surface spreading of liquid or culture samples uniformly over the surface of solid media. Shaped like a hockey stick, it is recommended for single use practices or sterile techniques.
Brand: Technical Service Consultants™ TS/30-B
Additional Details : Weight : 1.50000kg
Description
- Polystyrene
- Three year shelf life
- Ideal for bacterial counts
- L shaped plastic (firm) peel pouch
- Hockey Stick Shape
Specifications
Sterile | |
Spreader | |
Polystyrene |
L-Shape | |
500 Pk. |