Learn More
Nitritex™ Polyurethane Tacky Mat
Keep dirt, dust and bacteria at bay in controlled laboratory environments with the Nitritex™ Polyurethane Tacky Mat.
Brand: Nitritex™ STB2645
1345.63 SEK valid until 2025-05-02
Use promo code "24151" to get your promotional price.
This item is not returnable.
View return policy
Description
- Matting for use in controlled laboratory environments
- Rectangular
- Polyurethane
- White or blue
Specifications
ISO 9001:2015 | |
26 x 45 in. | |
Blue | |
Polyurethane |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
230316 | Certificate of Compliance | 07/03/2025 | STB2645 |
230472 | Certificate of Compliance | 21/08/2024 | STB2645 |
230102 | Certificate of Compliance | 09/07/2024 | STB2645 |
21-1025 | Certificate of Compliance | 31/08/2023 | STB2645 |
230590 | Certificate of Analysis | 30/05/2024 | STB2645 |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
New Offer Save Up to 92%
Product Content Correction
Product Title
Nitritex™ Polyurethane Tacky Mat > Size: 26 x 45in., Blue
Spot an opportunity for improvement?Share a Content Correction