Learn More
Kautex™ PVC Wide Neck Square Container, Transparent, Without Closure
Fill and clean these wide neck PVC containers with ease due to the roomy neck configuration. Supplied without closure.
Brand:  Kautex™ 2000070598
Additional Details : Weight : 0.02790kg
 View more versions of this product
Product Code. 10300721
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Member of Series 310 container family
  • Closure available separately
Specifications
Specifications
500 mL
54 mm
PVC
162/Cs.
Square
Wide Neck Container
Transparent
Packaging, Dispensing, Sending Samples, Storage, Mailing
162
No
108 mm
Closure, Foam Insert