Learn More
Kautex™ PVC Wide Neck Square Container, Transparent, Without Closure
Fill and clean these wide neck PVC containers with ease due to the roomy neck configuration. Supplied without closure.
Brand: Kautex™ 2000084422
Additional Details : Weight : 4.15800kg
Description
- Member of Series 310 container family
- Closure available separately
Specifications
500 mL | |
54 mm | |
PVC | |
126/Cs. | |
Square | |
Wide Neck Container |
Transparent | |
Packaging, Dispensing, Sending Samples, Storage, Mailing | |
126 | |
No | |
118.5 mm | |
Closure |