Learn More
Tocris Bioscience™ Pomalidomide 4'-alkylC2-amine
Pomalidomide-alkylC2-amine
Brand:  Tocris Bioscience™ 6851/25
2181.37 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18706766
  • 2765.00 SEK / 25mg
Explore more special offers
Description
Description
Pomalidomide 4'-alkylC2-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC2 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Pomalidomide - linker 7
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
4-[(2-Aminoethyl)amino]-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione hydrochloride
25 mg
352.78
Pomalidomide - linker 7
C15H16N4O4.HCl
95%