Learn More
Potassium 4-methoxyphenyltrifluoroborate, 95%, Thermo Scientific Chemicals
799.00 SEK - 2410.00 SEK
Chemical Identifiers
CAS | 192863-36-8 |
---|---|
Molecular Formula | C7H7BF3KO |
Molecular Weight (g/mol) | 214.036 |
MDL Number | MFCD04112717 |
InChI Key | XNYMCUFKZHRYID-UHFFFAOYSA-N |
Synonym | potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide |
PubChem CID | 23683762 |
IUPAC Name | potassium;trifluoro-(4-methoxyphenyl)boranuide |
SMILES | [B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357179
|
Thermo Scientific Alfa Aesar
H27384.03 |
1 g |
799.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367179
|
Thermo Scientific Alfa Aesar
H27384.06 |
5 g |
2410.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
192863-36-8 | |
214.036 | |
XNYMCUFKZHRYID-UHFFFAOYSA-N | |
23683762 | |
[B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+] |
C7H7BF3KO | |
MFCD04112717 | |
potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide | |
potassium;trifluoro-(4-methoxyphenyl)boranuide |
Specifications
192863-36-8 | |
C7H7BF3KO | |
potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide | |
[B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+] | |
214.036 | |
214.04 | |
Potassium 4-methoxyphenyltrifluoroborate |
>300°C | |
MFCD04112717 | |
XNYMCUFKZHRYID-UHFFFAOYSA-N | |
potassium;trifluoro-(4-methoxyphenyl)boranuide | |
23683762 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.