Learn More
Potassium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
441.00 SEK - 5925.00 SEK
Chemical Identifiers
CAS | 90076-67-8 |
---|---|
Molecular Formula | C2F6KNO4S2 |
Molecular Weight (g/mol) | 319.234 |
MDL Number | MFCD06200829 |
InChI Key | KVFIZLDWRFTUEM-UHFFFAOYSA-N |
Synonym | potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide |
PubChem CID | 11099217 |
IUPAC Name | potassium;bis(trifluoromethylsulfonyl)azanide |
SMILES | C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492797
|
Thermo Scientific Alfa Aesar
046909.MD |
250 mg |
441.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15472797
|
Thermo Scientific Alfa Aesar
046909.03 |
1 g |
1350.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15482797
|
Thermo Scientific Alfa Aesar
046909.06 |
5 g |
5925.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium bis(trifluoromethylsulfonyl)imide is used as Flavor & Fragrance intermediates. It is used as an intermediate in Industries such as agrochemical, dyestuff, pharmaceutical, syntheses material, chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
90076-67-8 | |
319.234 | |
KVFIZLDWRFTUEM-UHFFFAOYSA-N | |
11099217 | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+] |
C2F6KNO4S2 | |
MFCD06200829 | |
potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide | |
potassium;bis(trifluoromethylsulfonyl)azanide |
Specifications
90076-67-8 | |
C2F6KNO4S2 | |
MFCD06200829 | |
Moisture sensitive | |
It is soluble in water. 10mg/mL. | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+] | |
319.234 | |
319.24 | |
Potassium bis (trifluoromethylsulfonyl)imide |
198°C to 203°C | |
(CF3SO2)2NK | |
UN1759 | |
potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide | |
KVFIZLDWRFTUEM-UHFFFAOYSA-N | |
potassium;bis(trifluoromethylsulfonyl)azanide | |
11099217 | |
Solid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.