missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Solution (Conductivity Standard), 100.000μS/25°C, Chem Lab™
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Odor | Odorless |
Physical Form | Liquid |
Color | Colorless |
pH | ±5 |
Solubility | Soluble |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Chloride Solution (Conductivity Standard), 100.000μS/25°C, Chem Lab™ >
Spot an opportunity for improvement?Share a Content Correction