Learn More
Honeywell Fluka 1LT Potassium dichromate solution volumetric, 0.02 M K2Cr2O7 , for COD determination
Brand: Honeywell Fluka 34631-1L
1LT Potassium dichromate solution volumetric, 0.02 M K2Cr2O7 , for COD determination
Additional Details : CAS Number : 7778-50-9 Weight : 1.01000kg