missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydrogen phthalate, 99+%, Thermo Scientific Chemicals
1289.88 SEK - 3124.11 SEK
Chemical Identifiers
CAS | 877-24-7 |
---|---|
Molecular Formula | C8H5KO4 |
Molecular Weight (g/mol) | 204.22 |
MDL Number | MFCD00013070 |
InChI Key | IWZKICVEHNUQTL-UHFFFAOYSA-M |
Synonym | potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic |
PubChem CID | 23676735 |
IUPAC Name | potassium;2-carboxybenzoate |
SMILES | C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10516412
|
Thermo Scientific Acros
417955000 |
500 g | Glass bottle |
1289.88 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646324
|
Thermo Scientific Acros
417950025 |
2.5 kg | Plastic bottle |
3124.11 SEK
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
877-24-7 | |
204.22 | |
IWZKICVEHNUQTL-UHFFFAOYSA-M | |
23676735 | |
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
C8H5KO4 | |
MFCD00013070 | |
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic | |
potassium;2-carboxybenzoate |
Specifications
877-24-7 | |
Red | |
Authentic | |
C8H5KO4 | |
MFCD00013070 | |
15, 7733 | |
Solubility in water: 80g/L (20°C). Other solubilities: 330g/L water (100°C), slightly soluble in alcohol | |
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
204.22 | |
204.22 | |
Crystalline Powder |
295.0°C to 300.0°C | |
4 to 4.02 (0.05M soln. at 25°C) | |
99+% | |
2-(HO2C)C6H4CO2K | |
09, 791 | |
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic | |
IWZKICVEHNUQTL-UHFFFAOYSA-M | |
potassium;2-carboxybenzoate | |
23676735 | |
99+% | |
Potassium hydrogen phthalate, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 212-889-4
missing translation for 'rtecsNumber' : CZ4326000
missing translation for 'tsca' : TSCA
RUO – Research Use Only