Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Signal Generation for UF110plus
Potential-free contact for signal generation, controlled by program segment, for free-selectable functions to be activated.
Brand:  Memmert™ UF110plusH72
Product Code. 17518985
  • 6785.00 SEK / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • For signal generation
  • For free-selectable functions to be activated (e.g. activation of audible and visual signals, exhaust motors, fans, stirrers, etc.)
  • Only for units with TwinDISPLAY
  • Max. 2 contacts on 1-phase appliances
  • Max. 4 contacts on 3-phase appliances
Specifications
Specifications
UF110plus Oven
Socket to NAMUR NE 28
2 A
Potential-free Contact
24 V
Product Title
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Signal Generation for UF110plus > For Use With: UF110plus