Learn More
PPP4R2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22676-1-AP-150UL
 View more versions of this product
Product Code. 16822045
  • 4652.90 SEK / 150µL
Explore more special offers
Description
Description
The protein encoded by this gene is a regulatory subunit of the serine/threonine-protein phosphatase 4 complex. In addition to being required for efficient DNA double strand break repair, this complex plays a role in organization of microtubules at centrosomes and processing of spliceosomal snRNPs. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
PPP4R2
Polyclonal
Unconjugated
PPP4R2
PP4R2, PPP4R2
Rabbit
Antigen Affinity Chromatography
RUO
151987
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NY27
PPP4R2
PPP4R2 Fusion Protein Ag18486
150 μL
Primary
Human
Antibody
IgG
Product Title
PPP4R2 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated