missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP4R2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22676-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a regulatory subunit of the serine/threonine-protein phosphatase 4 complex. In addition to being required for efficient DNA double strand break repair, this complex plays a role in organization of microtubules at centrosomes and processing of spliceosomal snRNPs. Several transcript variants encoding different isoforms have been found for this gene.Specifications
PPP4R2 | |
Polyclonal | |
Unconjugated | |
PPP4R2 | |
PP4R2, PPP4R2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
151987 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NY27 | |
PPP4R2 | |
PPP4R2 Fusion Protein Ag18486 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPP4R2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction