missing translation for 'onlineSavingsMsg'
Learn More

Brady™ PR Plus Series Label Printer 9 to 9 Pin Cable

Cable accessory for PR Plus series label printers.

Brand:  Brady™ 360017

Product Code. 11513205

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Accessory for BradyPrinter™ model BP-PR Plus series.

Specifications

Specifications

White
9.8 ft.
2011/65/EU and 2015/863
BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer
3 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers