missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Praziquantel D11, MedChemExpress
MedChemExpress Praziquantel D11 is the deuterium labeled Praziquantel, which is an anthelmintic.
POA
Specifications
Chemical Name or Material | Praziquantel D11 |
---|---|
CAS | 1246343-36-1 |
Purity Grade Notes | Research |
Molecular Formula | C19H13D11N2O2 |
SMILES | O=C1N2CCC3=C(C=CC=C3)C2CN(C(C4([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C4([2H])[2H])=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16420848
|
Medchem Express
HY-B0244S |
5 mg |
POA
5mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16408438
|
Medchem Express
HY-B0244S |
10 mg |
POA
10mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Praziquantel D11 is the deuterium labeled Praziquantel, which is an anthelmintic.Specifications
Praziquantel D11 | |
Research | |
O=C1N2CCC3=C(C=CC=C3)C2CN(C(C4([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C4([2H])[2H])=O)C1 | |
323.47 | |
COVID-19-immunoregulation |
1246343-36-1 | |
C19H13D11N2O2 | |
323.47 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Please store the product under the recommended conditions in the Certificate of Analysis.
Recommended Storage : Please store the product under the recommended conditions in the Certificate of Analysis.
Research purposes only