Learn More
Kern Sohn Precision Balance EG
The classic balance with robust tuning fork measuring system.
Brand: Kern Sohn EG 4200-2NM
This item is not returnable.
View return policy
Description
- Internal adjustment in the case of a change in temperature and time-controlled at defined intervals, guarantees high degree of accuracy and makes the balance independent of its location of use
- Stable temperature behavior
- Short stabilization time
- Shock proof construction
- High corner load performance
- GLP/ISO record keeping of weight values
- Totalizing of pieces when counting
- Protective working cover included with delivery
Specifications
4200 g | |
0.02 g | |
180 mm | |
230 V, 50 Hz |
0.01 g | |
Precision Balance | |
160 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.