Learn More
Kern Sohn Precision Balance PLJ
High-quality precision scale with comfortable graphic display and enormous weighing range.
10695.42 SEK - 23642.51 SEK
Specifications
Type | Precision Balance |
---|---|
Electrical Requirements | Universal Mains Adapter |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
17311733
|
Kern Sohn
PLJ 1200-3A |
1200 g |
17225.26 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17321733
|
Kern Sohn
PLJ 2000-3A |
2100 g |
23642.51 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17341733
|
Kern Sohn
PLJ 4200-2F |
4200 g |
10695.42 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17331733
|
Kern Sohn
PLJ 420-3F |
420 g |
12271.59 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17351733
|
Kern Sohn
PLJ 6200-2A |
6200 g |
16662.34 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17361733
|
Kern Sohn
PLJ 6200-2AM |
6200 g (0.1 g verification) |
14973.59 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17371733
|
Kern Sohn
PLJ 720-3A |
720 g |
16774.92 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17381733
|
Kern Sohn
PLJ 720-3AM |
720 g (0.01 g verification) |
15198.75 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Convenient recipe-weighing: with the recipe database, in which up to 99 recipes can be stored, each with up to 20 recipe ingredients with name and target value
- Rapid and efficient operation thanks to the graphics display
- Simple, clear user interface on the display in the following languages: German, English, French, Italian, Spanish, Portuguese
- Automatic internal adjustment, guarantees high degree of accuracy and makes the balance independent of its location of use
- Dosage aid: High stability mode and other filter settings can be selected
- Internal memory for complete recipes with name and target value of the recipe ingredients. User guidance through display
- Protective working cover included with delivery
Specifications
Precision Balance | |
Universal Mains Adapter |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title