Learn More
Thermo Scientific™ Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Brand: Thermo Scientific™ 09.4000
Additional Details : Weight : 4.71600kg
Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.Specifications
Double cartridge pretreatment system (includes 5µm filter with carbon cartridge and a hardness stabilizer cartridge) | |
LabTower TII, Pacific RO, Pacific TII | |
Pretreatment Cartridge |