Learn More
Invitrogen™ PRIMER 5' AMINE 10 UMOL
Custom Primer
Brand:  Invitrogen™ 10715043
2332.50 SEK valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10759004
  • 3110.00 SEK / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5' Amine Primer, 10 nmol
5' Amine Primer
5' Labeled
10 nmol