Learn More
Invitrogen™ PRIMER PAGE 200 NMOL
Custom Primer
Brand:  Invitrogen™ 10916021
Product Code. 10367832
  • 410.00 SEK / Each
Explore more special offers
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Page Primer, 200 nmol
Page Primer
200 nmol
PAGE