missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pro-Brain Natriuretic Peptide Monoclonal Antibody (24E11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA120631
This item is not returnable.
View return policy
Description
MA1-20631 detects proBNP in human samples.
In cardiac tissue brain natriuretic peptide (BNP) is synthesized as 134 amino acid precursor (prepro-BNP), which is cleaved by proteases to form a 26 aa "e;signal"e; peptide and a 108 aa pro-BNP. Proteolytic digestion of pro-BNP results in formation of 76 aa amino-terminal NT-proBNP and biologically active 32 aa BNP hormone molecule. Both proBNP and NTproBNP circulate in human plasma and have been proposed as markers for early diagnosis of left ventricular dysfunction as well as prognostic markers of possible cardiac complications at patients with heart failure.Specifications
pro-Brain Natriuretic Peptide | |
Monoclonal | |
5.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P16860 | |
NPPB | |
Synthetic peptide corresponding to residues R(61) G H R K M V L Y T L R A P R(76) of human proBNP. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
24E11 | |
Unconjugated | |
NPPB | |
5 kDa cardiac natriuretic peptide; AA408272; BNF; BNP; BNP(1-28); BNP(1-29); BNP(1-30); BNP(1-32); BNP(2-31); BNP(3-29); BNP(3-30); BNP(3-32); BNP(4-27); BNP(4-29); BNP(4-30); BNP(4-31); BNP(4-32); BNP(5-29); BNP(5-31); BNP(5-32); BNP32; BNP-34; Brain natriuretic factor; brain natriuretic peptide; Brain natriuretic peptide 32; Brain natriuretic peptide 45; brain type natriuretic peptide; gamma-brain natriuretic peptide; Iso-ANP; natriuretic peptide B; natriuretic peptide precursor B; natriuretic peptide precursor type B; natriuretic peptide type B; natriuretic peptides B; natriuretic protein; Nppb; NTproBNP; type-B natriuretic factor | |
Mouse | |
Protein A | |
RUO | |
4879 | |
4° C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction