Learn More
Thermo Scientific™ Pro-Detect™ Rapid Human Fc Assay Kit
Pro-Detect™ Rapid Protein Fc Assay Kit is a single-use, dipstick lateral flow kit for quick, easy detection of human, rabbit or mouse Fc-tagged proteins in protein expression and purification systems.
Brand: Thermo Scientific™ A38504
Additional Details : Weight : 0.22680kg
Includes: 10 assays
Description
- Lateral flow assay uses membrane-based strips coated with dye-bound conjugated antibodies specific to the Fc tag to provide a visual, color readout for detection within ten minutes
- Detect tagged proteins directly from cell culture media (supernatant) or cell lysates without any special instrumentation or handling.
- Easy to adopt—lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
- Simple, fast method—simply dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
- Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentrations
- Specific—assay does not cross-react with common lysis and protein preparation reagents
- Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
- Long shelf life—stable for at least one year at 4°C
Specifications
• 10 Rapid Assay Strips • Sample Diluent, 15 mL Sufficient for 10 assay determinations Store at 2–8°C. |
|
Lateral Flow Rapid Assay | |
Pro-Detect™ | |
Cell Culture Supernatant, Cell Lysates | |
10 assays | |
Lateral Flow Rapid Assay | |
Wet Ice |
Human Fc-tagged proteins | |
Chromogenic | |
Assay | |
Antibody | |
Human | |
10 Assays |
For Research Use Only. Not for use in diagnostic procedures.