missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Procalcitonin Antibody (38F11), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-79065AF532
This item is not returnable.
View return policy
Description
Procalcitonin Monoclonal antibody specifically detects Procalcitonin in Human samples. It is validated for Western Blot, ELISASpecifications
Procalcitonin | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
38F11 | |
Western Blot, Sandwich ELISA | |
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fragments of human Procalcitonin conjugated with carrier protein. | |
0.2 mL | |
Cancer, Cellular Markers, Neuroscience, Phospho Specific, Signal Transduction | |
796 | |
Store at 4C in the dark. | |
IgG1 |