missing translation for 'onlineSavingsMsg'
Learn More

Systec™ D-Series Horizontal Benchtop Autoclave, Systec DE

Sterilize solids, liquids and hazardous biological substances with these autoclaves. Systec™ D-Series Horizontal Benchtop Autoclaves feature a compact and space-saving design. For basic laboratory applications and media sterilization.

Brand:  Systec™ 1650


Additional Details : Weight : 190.00000kg

Product Code. 12092959

  • POA / Each
Estimated Shipment: 05-09-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

Standard Features
  • Integrated, separate steam generator
  • Internal heating elements within the autoclave chamber
  • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
  • Temperature and pressure range 140°C, 4 bar
  • LCD display and fully automatic microprocessor control
  • Number of sterilization programs: Up to 25
  • Code-secured access rights for changing parameters and further safety-relevant intervention
  • Internal memory for storing up to 500 sterilization cycles
  • Timer for starting programs
  • Autofill: automatic demineralized water feed for steam generation
  • Flexible PT-100 temperature sensor
  • Temperature holding function for liquids after program finish
  • Special program for Durham tubes
  • Calculation of F0 value
  • Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
  • Water-cooled steam exhaust, thermostatically controlled
  • Programmable automatic door-opening on completion of program
  • RS-232 and RS-485 interfaces for external data transmission (network-compatible)
Available options
  • Extension of temperature and pressure ranges to 150°C / 5 bar (from chamber volume 65 liters and more)
Options for process optimization
  • Rapid cooling for efficient and safe cooling of liquids
  • Exhaust filtration (including condensate inactivation) for safe sterilization of hazardous biological substances
Options for documentation
  • Integrated printer for batch documentation
  • PC software for comprehensive documentation
  • Documentation SD: data storage on SD card for up to 10,000 sterilization cycles and transmission of data to a PC
Specifications

Specifications

1050 mm
160 L (Total), 150 L (Nominal)
500 mm
730 mm
D-Series Benchtop Autoclave
LCD display and fully automatic microprocessor control
4 bar
Horizontal benchtop
750 mm
Basic laboratory applications and media sterilization
DE-150
140°C
380 to 400 V 50/60 Hz 16 A 3-phase plus neutral
850 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers