Learn More
Fisherbrand™ MultiSUB Midi 96 Horizontal Gel System
Fisherbrand™ MultiSUB Midi 96 Horizontal Gel System, 96-well comb block and specialised tray replicates the format and well-spacing of microplates and thermal cycler blocks.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
Product Code. 12104011
  • 4890.00 SEK / Each
Estimated Shipment: 05-09-2024
to see stock.
Add to basket
Description
Description
  • Designed for loading of DNA samples from multi-well plates
  • Ideal for high throughput electrophoresis of PCR products or samples containing a small number of DNA bands
  • Average run-time is just 15 to 30 minutes
  • Direct microplate format for easy lane identification
  • Multichannel pipette compatible combs for speed loading
  • Stretch versions available with extended 3.6cm separation distance to double capacity
Specifications
Specifications
Horizontal Gel Electrophoresis System
Multi Sub Midi96 STRETCH, UV tray, Comb block with 12 x 8 sample, 1mm thick combs, casting dams