missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ ProfiLine™ pH 3310 Portable pH Meter
Portable, waterproof and robust meters for the determination of the parameters pH, ORP, dissolved oxygen, conductivity and BSB
Brand: WTW™ 2AA310
7958.04 SEK valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
The pH 3310 is an elegant combination of portable meter and data logger for all those who want to save measuring series automatically and keep processing them on a computer. The meter includes a built-in data memory for 5000 GLP-compliant entries with an interval-controlled logger. All data can conveniently be transmitted to a computer via the USB interface. The CMC function visualizes the optimum measuring range and thus supports correct measurements. The graphic display with clear text menus allows the comfortable and safe operation of the meter.Specifications
Data logger and USB interface, short instruction manual, CD-ROM and batteries, cable | |
±0.3, ±1 | |
For professional field measurements requiring documentation and data transfer | |
±0.005 | |
±0.3, ±1mV | |
LCD | |
CE | |
80 W x 180 H mm | |
3 Years | |
4 x 1.5V AA or 4 x 1.2V NiMH rechargeable | |
-5°C to +105°C | |
Portable pH meter | |
80 W mm | |
180 mm |
±0.1°C | |
±0.1, ±0.01, ±0.005 | |
-2 to 19.999 | |
-1200 to 1200, -2500 to 2500 | |
1, 2, 3, 4, 5 Calibration WTW Technical Buffer, DIN/NIST buffers plus 20 additional buffer set | |
Manual 500/5000 automatic | |
IP-66/67 | |
300 g | |
pH 3310 | |
All values ±1 digit | |
AA, NiMH | |
1000 hr. | |
80 mm | |
55 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction