missing translation for 'onlineSavingsMsg'
Learn More
Propargyltriphenylphosphonium bromide, 97%, Thermo Scientific™ Product Code.: 11465073

Propargyltriphenylphosphonium bromide, 97%, Thermo Scientific™

Product Code. 11465073
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11465073

Brand: Thermo Scientific Alfa Aesar A12753.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Propargyltriphenylphosphonium bromide can be used in the alcylation of amines. It is a commonly used as a Wittig reagent.

Solubility
Soluble in methanol.

Notes
Hygroscopic. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2091-46-5
Molecular Formula C21H18BrP
Molecular Weight (g/mol) 381.25
MDL Number MFCD00011811
InChI Key AFZDAWIXETXKRE-UHFFFAOYSA-M
Synonym triphenylpropargylphosphonium bromide, propargyltriphenylphosphonium bromide, prop-2-ynyltriphenylphosphonium bromide, triphenyl prop-2-yn-1-yl phosphonium bromide, propargyl triphenylphosphonium bromide, acmc-1cbag, propargyltriphenylphosphoniumbromide, triphenylpropargylphosphoniumbromide, triphenylprop-2-ynylphosphine, bromide, triphenyl-2-propynylphosphonium bromideShow MoreShow Less
PubChem CID 2724214
SMILES [Br-].C#CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 177°C to 182°C
Odor Odorless
Linear Formula HC≡CCH2P(C6H5)3Br
Quantity 25 g
Beilstein 3579775
Sensitivity Hygroscopic
Solubility Information Soluble in methanol.
IUPAC Name triphenyl(prop-2-yn-1-yl)phosphanium bromide
Formula Weight 381.26
Percent Purity 97%
Chemical Name or Material Propargyltriphenylphosphonium bromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD4659A Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Propargyltriphenylphosphonium bromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.