missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Kinase A Regulatory Subunit II alpha Rabbit anti-Human, Clone: 9C6B3, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | Protein Kinase A Regulatory Subunit II alpha |
---|---|
Clone | 9C6B3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18349213
|
Bio-Techne
NBP3-16292-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18321904
|
Bio-Techne
NBP3-16292-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein Kinase A Regulatory Subunit II alpha Monoclonal antibody specifically detects Protein Kinase A Regulatory Subunit II alpha in Human samples. It is validated for Western BlotSpecifications
Protein Kinase A Regulatory Subunit II alpha | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology, Cytoskeleton Markers, Endocrinology, GPCR, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
IgG | |
Affinity purified |
9C6B3 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Recombinant fusion protein containing a sequence corresponding to amino acids 312-404 of human Protein Kinase A Regulatory Subunit II alpha (P13861). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title