missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Phosphatase 1C gamma Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
2220.00 SEK - 4555.00 SEK
Specifications
Antigen | Protein Phosphatase 1C gamma |
---|---|
Applications | ChIP Assay, Immunohistochemistry, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18327196
|
Bio-Techne
NBP3-03445-20UL |
20 μL |
2220.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323215
|
Bio-Techne
NBP3-03445-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein Phosphatase 1C gamma Polyclonal specifically detects Protein Phosphatase 1C gamma in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
Protein Phosphatase 1C gamma | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 3.1.3.16, PP-1G, PP1gamma, PPP1G, protein phosphatase 1, catalytic subunit, gamma isoform, protein phosphatase 1, catalytic subunit, gamma isozyme, Protein phosphatase 1C catalytic subunit, serine/threonine phosphatase 1 gamma, serine/threonine-protein phosphatase PP1-gamma catalytic subunit | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human Protein Phosphatase 1C gamma (NP_002701.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
ChIP Assay, Immunohistochemistry, KnockDown | |
Unconjugated | |
Rabbit | |
Cancer, Lipid and Metabolism, Protein Phosphatase, Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
5501 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title