missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSA Monoclonal Antibody (5A11E9,5A11E2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515376
This item is not returnable.
View return policy
Description
MA5-15376 targets PSA (KLK3) in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human samples. The MA5-15376 immunogen is purified recombinant fragment of KLK3 (aa26-251) expressed in E. Coli. <br>.
PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and is abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.
Specifications
PSA | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P07288 | |
KLK3 | |
Purified recombinant fragment of KLK3 (aa26-251) expressed in E. Coli. | |
RUO | |
354 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
5A11E9,5A11E2, 5A11E2 | |
Unconjugated | |
KLK3 | |
Antigen; antigen, prostate specific; APS; Fletcher factor; gamma-seminoprotein; hK3; Kal3; Kal-3; Kallikrein 3; kallikrein 3, plasma; kallikrein B, plasma 1; kallikrein related peptidase 3; kallikrein-3; Kallikrein-3 (KLK3); kallikrein-related peptidase 3; Kalp1; KALP15; Kininogenin; KLK2A1; Klk3; KLK-3; Klkb1; P-30 antigen; Pk; Plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; prostate specific antigen; prostate-specific (APS); prostate-specific antigen; PSA; Semenogelase; seminin | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PSA Monoclonal Antibody (5A11E9,5A11E2), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction