missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAPL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558411
This item is not returnable.
View return policy
Description
Immunogen sequence: SITKECIILV DTYSPSLVQL VAKITPEKVC KFIRLCGNRR RARAVHDAYA IVPSPEWDAE NQGSFCNGCK RLLTVSSHN Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 61%.
May activate the lysosomal degradation of sphingolipids.Specifications
PSAPL1 | |
Polyclonal | |
Unconjugated | |
PSAPL1 | |
Proactivator polypeptide-like 1; prosaposin-like 1 (gene/pseudogene); prosaposin-like protein 1; PSAPL1; Saposin A-like; Saposin B-like; Saposin B-Val-like; Saposin C-like; Saposin D-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
768239 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NUJ1 | |
PSAPL1 | |
Recombinant protein corresponding to Human PSAPL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction