Learn More
Tocris Bioscience™ PSB 1114
Potent, selective P2Y2 agonist
Brand: Tocris Bioscience™ 4333/500U
2829.53 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
PSB 1114 is a potent, selective P2Y2 agonist (EC50 = 0.134 μM). Displays >60-fold selectivity versus P2Y4 and P2Y6 receptors.Specifications
4-Thiouridine-5'-O-(β,γ-difluoromethylene)triphosphate tetrasodium salt | |
500 μg | |
C10H11F2N2Na4O13P3S |
1657025-60-9 | |
P2Y Receptor Agonists | |
98% |