missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSD93 Rabbit anti-Human, Mouse, Rat, Clone: 0G4O6, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | PSD93 |
---|---|
Clone | 0G4O6 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18312146
|
Bio-Techne
NBP3-15849-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323465
|
Bio-Techne
NBP3-15849-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSD93 Monoclonal antibody specifically detects PSD93 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PSD93 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1740 | |
IgG | |
Affinity purified |
0G4O6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Chapsyn-110, discs, large homolog 2 (Drosophila), discs, large homolog 2, chapsyn-110, discs, large homolog 2, chapsyn-110 (Drosophila), disks large homolog 2, DKFZp781D1854, DKFZp781E0954, FLJ37266, MGC131811, Postsynaptic density protein PSD-93, PSD-93, PSD93,110kDa | |
A synthetic peptide corresponding to a sequence within amino acids 600-700 of human PSD93 (Q15700). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title