missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMA1 Rabbit anti-Human, Mouse, Rat, Clone: 5X3A6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16190-100UL
This item is not returnable.
View return policy
Description
PSMA1 Monoclonal antibody specifically detects PSMA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PSMA1 | |
Monoclonal | |
Unconjugated | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
5X3A6 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
A synthetic peptide corresponding to a sequence within amino acids 164-263 of human PSMA1 (P25786). | |
100 μg | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5682 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction