Learn More
PSMG3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA554907
Product Code. 15792831
  • 4900.00 SEK / 100µL
Explore more special offers
Description
Description
Immunogen sequence: VVVTQFGKMG TLVSLEPSSV ASDVSKPVLT TKVLLGQDEP LIHVFAKNLV AFVSQEAGNR AVLLAVAVKD KSMEGLKALR EVIRVCQ Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.
PSMG3, also named as C7orf48 and PAC3, is a chaperone protein which promotes assembly of the 20S proteasome. It may cooperate with PSMG1-PSMG2 heterodimers to orchestrate the correct assembly of proteasomes.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Title
PSMG3 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated