Learn More
PSMG3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA554907
Product Code. 15792831
  • 4900.00 SEK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Immunogen sequence: VVVTQFGKMG TLVSLEPSSV ASDVSKPVLT TKVLLGQDEP LIHVFAKNLV AFVSQEAGNR AVLLAVAVKD KSMEGLKALR EVIRVCQ Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.
PSMG3, also named as C7orf48 and PAC3, is a chaperone protein which promotes assembly of the 20S proteasome. It may cooperate with PSMG1-PSMG2 heterodimers to orchestrate the correct assembly of proteasomes.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PSMG3 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated