Learn More
Fisherbrand™ PTFE Blade for Plain Shafts
Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals. Cowie Technology™ PTFE Blade for Plain Shafts fit all standard shafts for blades.
Brand:  Fisherbrand™ 002.1065.1
 View more versions of this product
Product Code. 16285851
  • 391.00 SEK / Pack of 10
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of PTFE
  • Totally inert and non-scratch
  • Available in square end and plain end format
Specifications
Specifications
PTFE Shaft Stirrer, Plain Shaft for Blades
25 mm
-200°C to +280°C
Blades for Plain Shafts
Stirring in Narrow Neck Vessel
1 Each
3 mm
65 mm