Learn More
Bohlender™ PTFE Point Tip Tweezers
Use these tweezers for a variety of applications. BOLA PTFE Tweezers feature universal chemical resistance.
Brand:  Bohlender™ H909-02
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10319331
  • 262.88 SEK / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Precast tweezers
  • PTFE construction
  • With pointed ends
  • Universal chemical resistance
Specifications
Specifications
Pointed
Precast tweezer
100 mm
Each
PTFE
Tweezers