Learn More
Cole-Parmer™ PTFE-Coated Vacuum Pump
Wetted parts are coated with PTFE to handle your toughest applications
Brand:  Cole-Parmer™ 79200-10
Product Code. 11775169
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Lightweight aluminum construction and compact size make pumps ideal for benchtop applications such as vacuum filtration.
  • Pumps with gauges and regulators give you more control of your vacuum/pressure applications.
  • Include suction cup feet and a 6-ft power cord with built-in switch (the 115 VAC models have grounded plug, 220 VAC and DC models have stripped ends).
Specifications
Specifications
178.3 mmHg
Permanent magnet
Port size: 1/8 in. NPT(F)
104°F
33.6 L/min. Max.
18.73 mm
1/10 hp
5 A
PTFE
40°C
120.7 mm