Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Brand: Fisherbrand™ 001.803
Description
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
- Supplied in reusable snap cap vials.
- For stirring in test tubes, small size beakers, or other containers.
- Autoclavable.
Specifications
Autoclavable | |
White | |
Containers & Vessels / Stirrer Hot Plates | |
3 mm | |
10/Pk. | |
PTFE Stirrer Bars Micro |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE encapsulated Alnico V magnet core | |
PTFE | |
3 mm | |
Cylinder |