missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Plain Stirrer Shaft
Fisherbrand™ PTFE Plain Stirrer Shafts have a PTFE encapulated stainless steel core with exposed end. They are a fully molded construction featuring a PTFE roto molded to a fully PTFE encapsulated steel reinforced drive shaft.
Brand: Fisherbrand™ 005.101050EX
Description
- PTFE with Steel Core
- 50mm Exposed End
- Chemically Inert
- Easy to clean
- No cross contamination
- Non scratch
- Use up to 280° C
Specifications
PTFE Plain Stirrer Shaft | |
500 x 10 mm | |
1 |
6.35 mm | |
500 mm | |
10mm |