Learn More
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer are a fully molded product with a retreat angle of 30°.
875.00 SEK - 1140.00 SEK
Product Code | Brand | Dimensions (L x W) | Diameter (Metric) Fan | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Diameter (Metric) Fan | Price | Quantity & Availability | ||||
16205921
|
Fisherbrand™
005.80850.300 |
300 x 8 mm | 50 mm |
875.00 SEK
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
16215921
|
Fisherbrand™
005.80850.400 |
300 x 8 mm | 75 mm |
900.00 SEK
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
16225921
|
Fisherbrand™
005.80875.300 |
400 x 8 mm | 50 mm |
995.00 SEK
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
16235921
|
Fisherbrand™
005.80875.400 |
400 x 8 mm | 75 mm |
1025.00 SEK
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
16245921
|
Fisherbrand™
005.81050.400 |
400 x 10 mm | 50 mm |
995.00 SEK
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
16255921
|
Fisherbrand™
005.81050.500 |
500 x 10 mm | 50 mm |
1020.00 SEK
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
16265921
|
Fisherbrand™
005.81075.400 |
400 x 10 mm | 75 mm |
1080.00 SEK
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
16275921
|
Fisherbrand™
005.81075.500 |
500 x 10 mm | 75 mm |
1140.00 SEK
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
Fisherbrand™ PTFE Retreat Curve Shaft Stirrer | |
1 |