Learn More
Bohlender™ PTFE Socket-GL Tube Fittings
Brand: Bohlender™ D570-32
This item is not returnable.
View return policy
Specifications
FDA Conform | |
PTFE | |
-200°C | |
Socket NS 29/32 | |
250°C |
Socket-GL Tube Fittings | |
For connection of hard-walled tubing | |
1 Each | |
GL 25 |