Learn More
Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 16 to 20 in. wide
GREENER_CHOICE
PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.
Brand:  Keystone Cleanroom Products 6STKRP1828SS
 View more versions of this product
Product Code. 30042266
  • 53080.00 SEK / Pack of 250
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
  • Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
  • Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
  • Fast and consistent for equipment preparation area
  • Resists tearing and punctures
  • Superior microbial barrier
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Made in the USA
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Autoclave Bag
COC
Clear, White
Steam Sterilization of Pharmaceutical Equipment
Non-sterile
Yes
121 to 125 C
28 in.
18 in.
Autoclavable
Safe Seal
Single-use
Tyvek
5.3 mil
18 x 28
5.3 mil
71.1 cm
45.7 cm