missing translation for 'onlineSavingsMsg'
Learn More

Purine, 97+%, Thermo Scientific Chemicals

Product Code. 15453669
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Purine is a nucleotide base for proteomics research. It is also a significant component of many biomolecules such as adenine triphosphate, guanine triphosphate, cyclic adenosine monophosphate, nicotinamide adenine dinucleotide and coenzyme. Further, it is useful in medicinal chemistry as a building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Purine is a nucleotide base for proteomics research. It is also a significant component of many biomolecules such as adenine triphosphate, guanine triphosphate, cyclic adenosine monophosphate, nicotinamide adenine dinucleotide and coenzyme. Further, it is useful in medicinal chemistry as a building block.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-73-0
Molecular Formula C5H4N4
Molecular Weight (g/mol) 120.115
MDL Number MFCD00079221
InChI Key KDCGOANMDULRCW-UHFFFAOYSA-N
Synonym purine, 9h-purine, 1h-purine, isopurine, beta-purine, 7h-imidazo 4,5-d pyrimidine, imidazo 4,5-d pyrimidine, 3,5,7-triazaindole, 9h-purine van, 3h-purine
PubChem CID 1044
ChEBI CHEBI:17258
IUPAC Name 7H-purine
SMILES C1=C2C(=NC=N1)N=CN2

Specifications

Melting Point 215°C to 217°C
Color White
Quantity 1 g
Beilstein 3200
Merck Index 14,7942
Solubility Information Soluble in water.
Formula Weight 120.11
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Purine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'