missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ PVC Drawer Organzer
Organize your 25mm diameter vials with the Vitlab™ PVC Drawer Organizer.
Brand: VITLAB™ 80952
This item is not returnable.
View return policy
Description
- White
- 9 lengthwise compartments opening at the front
- For vials with a diameter of 25 mm
Specifications
45 mm | |
PVC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction